Product Details

SNP ID
rs146558038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10929687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGCCCACTCGCTGGTGCAGGC[A/G]GAGGCCCCGAGATGAAACCCTGAGG
Phenotype
MIM: 603934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf52 PubMed Links

Gene Details

Gene
C19orf52
Gene Name
chromosome 19 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138358.2 843 Silent Mutation GCA,GCG A256A NP_612367.1
Gene
CARM1
Gene Name
coactivator associated arginine methyltransferase 1
There are no transcripts associated with this gene.

Gene
YIPF2
Gene Name
Yip1 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321439.1 843 Intron NP_001308368.1
NM_001321440.1 843 Intron NP_001308369.1
NM_024029.4 843 Intron NP_076934.1
XM_011528270.2 843 Intron XP_011526572.1
XM_011528271.2 843 Intron XP_011526573.1

View Full Product Details