Product Details

SNP ID
rs148085889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39391451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCATCCCAGCAGCAAGCTTCAG[C/T]GGCTTCCTCAGCTGCTGGTGTATCG
Phenotype
MIM: 612914 MIM: 610506
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED29 PubMed Links

Gene Details

Gene
MED29
Gene Name
mediator complex subunit 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317770.1 149 Missense Mutation GCG,GTG A31V NP_001304699.1
NM_017592.2 149 Missense Mutation GCG,GTG A31V NP_060062.1
Gene
PAF1
Gene Name
PAF1 homolog, Paf1/RNA polymerase II complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256826.1 149 Intron NP_001243755.1
NM_019088.3 149 Intron NP_061961.2
Gene
SAMD4B
Gene Name
sterile alpha motif domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303614.1 149 Intron NP_001290543.1
NM_018028.3 149 Intron NP_060498.2
XM_011527057.2 149 Intron XP_011525359.1
XM_011527060.2 149 Intron XP_011525362.1
XM_011527061.2 149 Intron XP_011525363.1
XM_011527062.2 149 Intron XP_011525364.1
XM_011527063.2 149 Intron XP_011525365.1
XM_011527064.2 149 Intron XP_011525366.1
XM_017026918.1 149 Intron XP_016882407.1
XM_017026919.1 149 Intron XP_016882408.1
XM_017026920.1 149 Intron XP_016882409.1
XM_017026921.1 149 Intron XP_016882410.1
XM_017026922.1 149 Intron XP_016882411.1
XM_017026923.1 149 Intron XP_016882412.1

View Full Product Details