Product Details

SNP ID
rs111381847
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:211576150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACAATCACTGAACCCAAGGCATC[C/T]CCAAGGACATGCAGAAAAACTCCAC
Phenotype
MIM: 609521
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC30A1 PubMed Links

Gene Details

Gene
SLC30A1
Gene Name
solute carrier family 30 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021194.2 907 Silent Mutation GGA,GGG G254G NP_067017.2

View Full Product Details