Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001319179.1 | 1801 | Missense Mutation | CAG,GAG | Q476E | NP_001306108.1 |
NM_001319180.1 | 1801 | Missense Mutation | CAG,GAG | Q421E | NP_001306109.1 |
NM_001319181.1 | 1801 | Intron | NP_001306110.1 | ||
NM_018284.2 | 1801 | Missense Mutation | CAG,GAG | Q555E | NP_060754.2 |
XM_005270743.2 | 1801 | Nonsense Mutation | CAG,GAG | Q476E | XP_005270800.1 |
XM_006710574.2 | 1801 | Nonsense Mutation | CAG,GAG | Q528E | XP_006710637.1 |
XM_011541233.1 | 1801 | Nonsense Mutation | CAG,GAG | Q476E | XP_011539535.1 |
XM_011541234.1 | 1801 | Nonsense Mutation | CAG,GAG | Q474E | XP_011539536.1 |
XM_017001013.1 | 1801 | Nonsense Mutation | CAG,GAG | Q421E | XP_016856502.1 |
XM_017001014.1 | 1801 | Missense Mutation | CAG,GAG | Q340E | XP_016856503.1 |