Product Details

SNP ID
rs112516583
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:54648856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTGAGAGCTGTTCTGGAGCCA[C/G]AATTAGCAGAACTTGGCCAAAATTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MROH7 PubMed Links

Gene Details

Gene
MROH7
Gene Name
maestro heat like repeat family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039464.3 Intron NP_001034553.2
NM_001291332.1 Intron NP_001278261.1
Gene
MROH7-TTC4
Gene Name
MROH7-TTC4 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details