Product Details

SNP ID
rs139370579
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:209941058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTTTTGAACTGAACAGATCGTT[C/T]ATATCTGTTAAGCATACTCTTATTA
Phenotype
MIM: 610949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYT14 PubMed Links

Gene Details

Gene
SYT14
Gene Name
synaptotagmin 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146261.2 Intron NP_001139733.1
NM_001146262.2 Intron NP_001139734.1
NM_001146264.2 Intron NP_001139736.1
NM_001256006.1 Intron NP_001242935.1
NM_153262.3 Intron NP_694994.2
XM_006711262.3 Intron XP_006711325.1
XM_017000931.1 Intron XP_016856420.1
XM_017000932.1 Intron XP_016856421.1
XM_017000933.1 Intron XP_016856422.1
XM_017000934.1 Intron XP_016856423.1
XM_017000935.1 Intron XP_016856424.1

View Full Product Details