Product Details

SNP ID
rs140022471
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:206685427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCATCGATGACTACAAGGTCAC[C/T]AGCCAGGTCCTGGGGCTGGGCATCA
Phenotype
MIM: 602006
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPKAPK2 PubMed Links

Gene Details

Gene
MAPKAPK2
Gene Name
mitogen-activated protein kinase-activated protein kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004759.4 408 Silent Mutation ACC,ACT T66T NP_004750.1
NM_032960.3 408 Silent Mutation ACC,ACT T66T NP_116584.2
XM_005273353.3 408 Silent Mutation ACC,ACT T66T XP_005273410.1
XM_017002810.1 408 Intron XP_016858299.1

View Full Product Details