Product Details
- SNP ID
-
rs141183652
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:3631573 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCTGCTGCGGGTCCCACTGAAAT[A/G]CGCGCACTGGGGACAGCTGCTCGAG
- Phenotype
-
MIM: 604266
MIM: 611460
MIM: 606040
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MEGF6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2760321] are located under a probe and SNP(s) [rs16823940] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MEGF6
- Gene Name
- multiple EGF like domains 6
There are no transcripts associated with this gene.
- Gene
- TPRG1L
- Gene Name
- tumor protein p63 regulated 1-like
There are no transcripts associated with this gene.
- Gene
- WRAP73
- Gene Name
- WD repeat containing, antisense to TP73
View Full Product Details