Product Details

SNP ID
rs141956734
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:48259808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTAAAATTTTCCCTGGACCATT[C/T]CTTTCTCTAGCAGAATTAATCATAT
Phenotype
MIM: 613947
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPATA6 PubMed Links

Gene Details

Gene
SPATA6
Gene Name
spermatogenesis associated 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286238.1 3341 Intron NP_001273167.1
NM_001286239.1 3341 Intron NP_001273168.1
NM_019073.3 3341 Intron NP_061946.1
XM_006710699.2 3341 UTR 3 XP_006710762.1
XM_006710700.2 3341 Intron XP_006710763.1
XM_006710701.3 3341 Intron XP_006710764.1
XM_011541606.1 3341 Intron XP_011539908.1
XM_011541607.2 3341 Intron XP_011539909.1
XM_011541608.1 3341 Intron XP_011539910.1
XM_011541609.2 3341 Intron XP_011539911.1
XM_017001509.1 3341 Intron XP_016856998.1
XM_017001510.1 3341 Intron XP_016856999.1
XM_017001511.1 3341 Intron XP_016857000.1
XM_017001512.1 3341 Intron XP_016857001.1
XM_017001513.1 3341 Intron XP_016857002.1
XM_017001514.1 3341 Intron XP_016857003.1
XM_017001515.1 3341 Intron XP_016857004.1

View Full Product Details