Product Details

SNP ID
rs144493145
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:246726971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTCGCTTGATGAAATGGCTAA[A/G]CAGGCAACAGTTGTCCTCAATTGCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCCPDH PubMed Links

Gene Details

Gene
SCCPDH
Gene Name
saccharopine dehydrogenase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016002.2 617 Silent Mutation AAA,AAG K90K NP_057086.2

View Full Product Details