Product Details

SNP ID
rs145060091
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:231347881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGGCATGGTCCAGAATTTTGT[A/G]AACATATGCATCGCATCAACAGCCT
Phenotype
MIM: 616086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPRTN PubMed Links

Gene Details

Gene
SPRTN
Gene Name
SprT-like N-terminal domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010984.3 854 Missense Mutation AAA,GAA K136E NP_001010984.1
NM_001261462.2 854 Intron NP_001248391.1
NM_032018.6 854 Missense Mutation AAA,GAA K136E NP_114407.3
XM_006711818.2 854 Intron XP_006711881.1
XM_011544289.2 854 Intron XP_011542591.1

View Full Product Details