Product Details

SNP ID
rs145155021
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:160367866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCTCCCAGCCCCAGCATGGGG[A/G]GTGGGGAGTTGGGGGCACTAAACAT
Phenotype
MIM: 605254 MIM: 162360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCSTN PubMed Links

Gene Details

Gene
NCSTN
Gene Name
nicastrin
There are no transcripts associated with this gene.

Gene
NHLH1
Gene Name
nescient helix-loop-helix 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005598.3 Intron NP_005589.1

View Full Product Details