Product Details

SNP ID
rs146391078
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:47094635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAAGATACTACTCAGAAAAGGC[A/G]CTGGGATTTTCTGGACATACTTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP4Z1 PubMed Links

Gene Details

Gene
CYP4Z1
Gene Name
cytochrome P450 family 4 subfamily Z member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178134.2 Intron NP_835235.1

View Full Product Details