Product Details

SNP ID
rs146391313
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.1:27669252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTACCCGCATTCTCACCTGCCTC[C/T]ACCCCACTGCAAGTGAAGAGCAGCA
Phenotype
MIM: 147572
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
IFI6 PubMed Links

Gene Details

Gene
IFI6
Gene Name
interferon alpha inducible protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002038.3 186 Silent Mutation GTA,GTG V21V NP_002029.3
NM_022872.2 186 Silent Mutation GTA,GTG V21V NP_075010.1
NM_022873.2 186 Silent Mutation GTA,GTG V21V NP_075011.1

View Full Product Details