Product Details

SNP ID
rs147315301
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:25225018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACTTACTGTTTTTCCAGATCTA[C/T]GACCATCCTGTCAATTTCCTCTGTG
Phenotype
MIM: 607090
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYF2 PubMed Links

Gene Details

Gene
SYF2
Gene Name
SYF2 pre-mRNA splicing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015484.4 479 Missense Mutation ATA,GTA I184V NP_056299.1
NM_207170.3 479 Missense Mutation ATA,GTA I142V NP_997053.1

View Full Product Details