Product Details

SNP ID
rs147803387
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:15443467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCAGAGCATGTGGAGCTGAGTGA[C/G]ACCATCCAGGTGGCCTGCCTGCCAG
Phenotype
MIM: 601405
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTRC PubMed Links

Gene Details

Gene
CTRC
Gene Name
chymotrypsin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007272.2 428 Missense Mutation GAC,GAG D135E NP_009203.2
XM_011540550.1 428 Missense Mutation GAC,GAG D135E XP_011538852.1

View Full Product Details