Product Details

SNP ID
rs148135133
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:246726953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGATATTGCTAATCCAGCCTC[A/G]CTTGATGAAATGGCTAAACAGGCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCCPDH PubMed Links

Gene Details

Gene
SCCPDH
Gene Name
saccharopine dehydrogenase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016002.2 599 Silent Mutation TCA,TCG S84S NP_057086.2

View Full Product Details