Product Details

SNP ID
rs148595354
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:15442518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAAGACGAAGAAGGATCCCTGT[C/T]TGTGGGTGTGGACACCATCCACGTC
Phenotype
MIM: 601405
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTRC PubMed Links

Gene Details

Gene
CTRC
Gene Name
chymotrypsin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007272.2 325 Missense Mutation TCT,TTT S101F NP_009203.2
XM_011540550.1 325 Missense Mutation TCT,TTT S101F XP_011538852.1

View Full Product Details