Product Details

SNP ID
rs148622513
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.1:112702644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCCCCACCAGGATGATGGGCAC[A/G]TTGGGGCAGAAGTGCTTCACCTCTG
Phenotype
MIM: 610742 MIM: 609957 MIM: 165380
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MOV10 PubMed Links
Additional Information
For this assay, SNP(s) [rs2245013] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MOV10
Gene Name
Mov10 RISC complex RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130079.2 660 Intron NP_001123551.1
NM_001286072.1 660 Intron NP_001273001.1
NM_001321324.1 660 Intron NP_001308253.1
NM_020963.4 660 Intron NP_066014.1
XM_005270869.4 660 Intron XP_005270926.1
XM_005270870.4 660 Intron XP_005270927.1
XM_017001318.1 660 Intron XP_016856807.1
XM_017001319.1 660 Intron XP_016856808.1
Gene
PPM1J
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1J
There are no transcripts associated with this gene.

Gene
RHOC
Gene Name
ras homolog family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042678.1 660 Silent Mutation AAC,AAT N109N NP_001036143.1
NM_001042679.1 660 Silent Mutation AAC,AAT N109N NP_001036144.1
NM_175744.4 660 Silent Mutation AAC,AAT N109N NP_786886.1

View Full Product Details