Product Details

SNP ID
rs149454614
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:27668535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGCTCTCCGAGCACTTTTTCTTA[C/T]CTGCAGGAGAGCAGACAAAGCGTAG
Phenotype
MIM: 147572
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFI6 PubMed Links

Gene Details

Gene
IFI6
Gene Name
interferon alpha inducible protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002038.3 206 Missense Mutation GAT,GGT D24G NP_002029.3
NM_022872.2 206 Missense Mutation GAT,GGT D28G NP_075010.1
NM_022873.2 206 Missense Mutation GAT,GGT D32G NP_075011.1

View Full Product Details