Product Details

SNP ID
rs149516397
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:3630978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACACGTTAGTGCACCGCTGCTA[C/T]GTGTGGCCGCCCAGCTGTCTGCAGG
Phenotype
MIM: 604266 MIM: 611460 MIM: 606040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEGF6 PubMed Links

Gene Details

Gene
MEGF6
Gene Name
multiple EGF like domains 6
There are no transcripts associated with this gene.

Gene
TPRG1L
Gene Name
tumor protein p63 regulated 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182752.3 1462 Intron NP_877429.2
Gene
WRAP73
Gene Name
WD repeat containing, antisense to TP73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017818.3 1462 Silent Mutation NP_060288.3
XM_005244754.1 1462 Silent Mutation XP_005244811.1
XM_017001387.1 1462 Silent Mutation XP_016856876.1

View Full Product Details