Product Details
- SNP ID
-
rs149036444
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:46350507 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCTGGAGCCCAGCCCCTTCAAGG[A/C]CTTGGGGCCACCATCACCTTGGGAT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SLC35C2
PubMed Links
Gene Details
- Gene
- SLC35C2
- Gene Name
- solute carrier family 35 member C2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001281457.1 |
1245 |
Missense Mutation |
GCC,TCC |
A215S |
NP_001268386.1 |
NM_001281458.1 |
1245 |
Missense Mutation |
GCC,TCC |
A358S |
NP_001268387.1 |
NM_001281459.1 |
1245 |
Missense Mutation |
GCC,TCC |
A194S |
NP_001268388.1 |
NM_001281460.1 |
1245 |
Missense Mutation |
GCC,TCC |
A329S |
NP_001268389.1 |
NM_015945.11 |
1245 |
Missense Mutation |
GCC,TCC |
A329S |
NP_057029.8 |
NM_173073.3 |
1245 |
Missense Mutation |
GCC,TCC |
A308S |
NP_775096.1 |
NM_173179.3 |
1245 |
Missense Mutation |
GCC,TCC |
A329S |
NP_775271.1 |
XM_011528831.1 |
1245 |
Missense Mutation |
GCC,TCC |
A329S |
XP_011527133.1 |
XM_011528832.1 |
1245 |
Missense Mutation |
GCC,TCC |
A329S |
XP_011527134.1 |
XM_011528833.1 |
1245 |
Missense Mutation |
GCC,TCC |
A329S |
XP_011527135.1 |
XM_011528834.1 |
1245 |
Missense Mutation |
GCC,TCC |
A328S |
XP_011527136.1 |
XM_011528835.1 |
1245 |
Missense Mutation |
GCC,TCC |
A291S |
XP_011527137.1 |
XM_011528836.1 |
1245 |
Missense Mutation |
GCC,TCC |
A215S |
XP_011527138.1 |
XM_011528837.1 |
1245 |
Missense Mutation |
GCC,TCC |
A215S |
XP_011527139.1 |
XM_011528838.1 |
1245 |
Missense Mutation |
GCC,TCC |
A177S |
XP_011527140.1 |
XM_017027856.1 |
1245 |
Missense Mutation |
GCC,TCC |
A329S |
XP_016883345.1 |
XM_017027857.1 |
1245 |
Missense Mutation |
GCC,TCC |
A329S |
XP_016883346.1 |
XM_017027858.1 |
1245 |
Missense Mutation |
GCC,TCC |
A329S |
XP_016883347.1 |
XM_017027859.1 |
1245 |
Missense Mutation |
GCC,TCC |
A329S |
XP_016883348.1 |
XM_017027860.1 |
1245 |
Missense Mutation |
GCC,TCC |
A329S |
XP_016883349.1 |
XM_017027861.1 |
1245 |
Missense Mutation |
GCC,TCC |
A308S |
XP_016883350.1 |
XM_017027862.1 |
1245 |
UTR 3 |
|
|
XP_016883351.1 |
View Full Product Details