Product Details

SNP ID
rs138723870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24938189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGGCATTGGAACACTGCTTCT[C/G]CTTGGGCACAGCCCTTTGGGGGCCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM211 PubMed Links

Gene Details

Gene
TMEM211
Gene Name
transmembrane protein 211
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001663.1 112 Silent Mutation AGC,AGG S29R NP_001001663.1
XM_011530094.2 112 Missense Mutation AGC,AGG S90R XP_011528396.1

View Full Product Details