Product Details

SNP ID
rs139006374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35299961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCTGGGGAACCCGTTCAGCTC[G/T]CCAGTGGGACAGCGCATCGGTGAGT
Phenotype
MIM: 604702 MIM: 604700
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HMGXB4 PubMed Links

Gene Details

Gene
HMGXB4
Gene Name
HMG-box containing 4
There are no transcripts associated with this gene.

Gene
TOM1
Gene Name
target of myb1 membrane trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135729.1 158 Intron NP_001129201.1
NM_001135730.1 158 Silent Mutation TCG,TCT S11S NP_001129202.1
NM_001135732.1 158 Silent Mutation TCG,TCT S11S NP_001129204.1
NM_005488.2 158 Silent Mutation TCG,TCT S11S NP_005479.1
XM_011529818.2 158 UTR 5 XP_011528120.1
XM_011529820.2 158 UTR 5 XP_011528122.1
XM_017028529.1 158 UTR 5 XP_016884018.1
XM_017028530.1 158 UTR 5 XP_016884019.1

View Full Product Details