Product Details

SNP ID
rs147385961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23059599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGTCTGCAGGGCCTTGCGGCCCT[C/T]GGGGGCCTCTGCCAGCATGGTAAGG
Phenotype
MIM: 605663
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSPH14 PubMed Links

Gene Details

Gene
RSPH14
Gene Name
radial spoke head 14 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014433.2 1403 Missense Mutation AAG,GAG K304E NP_055248.1
XM_011530149.2 1403 Missense Mutation AAG,GAG K354E XP_011528451.1
XM_011530150.1 1403 Missense Mutation CAA,CGA Q348R XP_011528452.1
XM_011530151.2 1403 Missense Mutation CAA,CGA Q348R XP_011528453.1
XM_011530152.2 1403 Missense Mutation CAA,CGA Q348R XP_011528454.1
XM_011530154.2 1403 Missense Mutation CAA,CGA Q280R XP_011528456.1
XM_011530155.2 1403 Missense Mutation CAA,CGA Q216R XP_011528457.1
XM_017028774.1 1403 Missense Mutation CAA,CGA Q348R XP_016884263.1

View Full Product Details