Product Details

SNP ID
rs140966980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241606241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGCTTGGCCTTTGTCTTTGAT[A/C]AGTCTGGAATTATTTTCAGAGACCT
Phenotype
MIM: 612533
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
THAP4 PubMed Links

Gene Details

Gene
THAP4
Gene Name
THAP domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164356.1 Intron NP_001157828.1
NM_015963.5 Intron NP_057047.4
XM_005247016.4 Intron XP_005247073.3
XM_011511291.2 Intron XP_011509593.1
XM_017004256.1 Intron XP_016859745.1

View Full Product Details