Product Details

SNP ID
rs141801834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189563614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGCAGCAATGACGCCTGCAAACA[C/G]CAGACTGACAGAGATTATGGGCACA
Phenotype
MIM: 604653
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC40A1 PubMed Links

Gene Details

Gene
SLC40A1
Gene Name
solute carrier family 40 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014585.5 1858 Missense Mutation CTG,GTG L458V NP_055400.1
XM_005246505.1 1858 Missense Mutation CTG,GTG L418V XP_005246562.1
XM_017003938.1 1858 Missense Mutation CTG,GTG L418V XP_016859427.1

View Full Product Details