Product Details
- SNP ID
-
rs142855358
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:70258194 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCGCGTCGTCGCGGAGCTCGTCTC[C/T]TCGCTGCTGGGGTTGTGGCTGTTGC
- Phenotype
-
MIM: 610995
MIM: 603518
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR1285-2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11556184] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR1285-2
- Gene Name
- microRNA 1285-2
There are no transcripts associated with this gene.
- Gene
- PCYOX1
- Gene Name
- prenylcysteine oxidase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016297.3 |
96 |
Silent Mutation |
TCC,TCT |
S10S |
NP_057381.3 |
- Gene
- TIA1
- Gene Name
- TIA1 cytotoxic granule-associated RNA binding protein
There are no transcripts associated with this gene.
View Full Product Details