Product Details

SNP ID
rs144292380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169128750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGGTTGGAGTTGGGTCTCTTCT[C/T]GAAGGAGGCTTAGGCTTAGCAGGGA
Phenotype
MIM: 600073
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP2 PubMed Links

Gene Details

Gene
LRP2
Gene Name
LDL receptor related protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004525.2 13966 Silent Mutation TCA,TCG S4627S NP_004516.2
XM_011511183.2 13966 Silent Mutation TCA,TCG S4584S XP_011509485.1
XM_011511184.2 13966 Silent Mutation TCA,TCG S3864S XP_011509486.1

View Full Product Details