Product Details

SNP ID
rs145520267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88126341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCCCTTCTGGATGAGCTCTTC[C/T]GGCAGACCTGGTGGAAACCGTCTGA
Phenotype
MIM: 134650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FABP1 PubMed Links

Gene Details

Gene
FABP1
Gene Name
fatty acid binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001443.2 189 Silent Mutation CCA,CCG P25P NP_001434.1

View Full Product Details