Product Details

SNP ID
rs149918549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20249388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGACAGCGGCTTTGTGTGGGTTT[C/T]AAAAAAATGAAACAAACACAAGGCG
Phenotype
MIM: 607205
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PUM2 PubMed Links

Gene Details

Gene
PUM2
Gene Name
pumilio RNA binding family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282752.1 5247 UTR 3 NP_001269681.1
NM_001282790.1 5247 UTR 3 NP_001269719.1
NM_001282791.1 5247 UTR 3 NP_001269720.1
NM_015317.2 5247 UTR 3 NP_056132.1
XM_005262607.1 5247 UTR 3 XP_005262664.1
XM_005262609.1 5247 UTR 3 XP_005262666.1
XM_005262610.1 5247 UTR 3 XP_005262667.1
XM_006711973.1 5247 UTR 3 XP_006712036.1
XM_006711974.2 5247 UTR 3 XP_006712037.1
XM_006711975.2 5247 UTR 3 XP_006712038.1
XM_011532719.1 5247 UTR 3 XP_011531021.1
XM_011532720.2 5247 UTR 3 XP_011531022.1
XM_011532721.2 5247 UTR 3 XP_011531023.1
XM_011532723.1 5247 UTR 3 XP_011531025.1
XM_011532724.2 5247 Intron XP_011531026.1
XM_011532725.2 5247 Intron XP_011531027.1
XM_017003701.1 5247 UTR 3 XP_016859190.1
XM_017003702.1 5247 UTR 3 XP_016859191.1
XM_017003703.1 5247 UTR 3 XP_016859192.1
XM_017003704.1 5247 UTR 3 XP_016859193.1
XM_017003705.1 5247 UTR 3 XP_016859194.1
XM_017003706.1 5247 UTR 3 XP_016859195.1
XM_017003707.1 5247 UTR 3 XP_016859196.1
XM_017003708.1 5247 UTR 3 XP_016859197.1
XM_017003709.1 5247 UTR 3 XP_016859198.1
XM_017003710.1 5247 UTR 3 XP_016859199.1
XM_017003711.1 5247 UTR 3 XP_016859200.1
XM_017003712.1 5247 UTR 3 XP_016859201.1
XM_017003713.1 5247 UTR 3 XP_016859202.1
XM_017003714.1 5247 UTR 3 XP_016859203.1
XM_017003715.1 5247 UTR 3 XP_016859204.1

View Full Product Details