Product Details

SNP ID
rs141976295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195733343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGTACCCAGAAGGTTCCCATGA[A/C]GGGCAGCATGTCCAAGCCCCTGACC
Phenotype
MIM: 610360
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MUC20 PubMed Links

Gene Details

Gene
MUC20
Gene Name
mucin 20, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282506.1 2381 UTR 3 NP_001269435.1
NM_001291833.1 2381 UTR 3 NP_001278762.1
NM_020790.1 2381 UTR 3 NP_065841.1
NM_152673.3 2381 UTR 3 NP_689886.3

View Full Product Details