Product Details

SNP ID
rs144813471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48626924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGTCCACAAAGGAGAGGGCACC[A/C]AGGTCCAGGATGAGGCTGTGGAAGT
Phenotype
MIM: 604264 MIM: 610068
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CELSR3 PubMed Links

Gene Details

Gene
CELSR3
Gene Name
cadherin EGF LAG seven-pass G-type receptor 3
There are no transcripts associated with this gene.

Gene
MIR6824
Gene Name
microRNA 6824
There are no transcripts associated with this gene.

Gene
SLC26A6
Gene Name
solute carrier family 26 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040454.1 2125 Silent Mutation NP_001035544.1
NM_001281732.1 2125 Silent Mutation NP_001268661.1
NM_001281733.1 2125 Silent Mutation NP_001268662.1
NM_022911.2 2125 Silent Mutation NP_075062.2
NM_134263.2 2125 Silent Mutation NP_599025.2
NM_134426.2 2125 Silent Mutation NP_602298.2
Gene
TMEM89
Gene Name
transmembrane protein 89
There are no transcripts associated with this gene.

View Full Product Details