Product Details

SNP ID
rs145087533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129976766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAGGCAAAAGAGAGGAGGAGGA[G/T]GAAGAGGGAGTTGAAGAAGAGGAAG
Phenotype
MIM: 613879
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRH PubMed Links

Gene Details

Gene
TRH
Gene Name
thyrotropin releasing hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007117.4 718 Missense Mutation GAG,GAT E93D NP_009048.1

View Full Product Details