Product Details

SNP ID
rs145660527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:197951168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTTTGTGTCTTAGGCTGTGGTC[C/T]AAGGCCATTTTTGCTGGCTATAAGC
Phenotype
MIM: 612477 MIM: 609380 MIM: 180468
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQCG PubMed Links

Gene Details

Gene
IQCG
Gene Name
IQ motif containing G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134435.2 94 Intron NP_001127907.1
NM_001323027.1 94 Intron NP_001309956.1
NM_001323028.1 94 Intron NP_001309957.1
NM_001323029.1 94 Intron NP_001309958.1
NM_001323030.1 94 Intron NP_001309959.1
NM_032263.4 94 Intron NP_115639.1
XM_017007316.1 94 Intron XP_016862805.1
XM_017007317.1 94 Intron XP_016862806.1
Gene
LMLN
Gene Name
leishmanolysin like peptidase
There are no transcripts associated with this gene.

Gene
RPL35A
Gene Name
ribosomal protein L35a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000996.2 94 Silent Mutation NP_000987.2
NM_001316311.1 94 Intron NP_001303240.1

View Full Product Details