Product Details

SNP ID
rs145868396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113004494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTTCTGTTATGATAGGGTGGCT[A/G]AGGAAAAGAAGGATGCGGTACATAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTPBP8 PubMed Links

Gene Details

Gene
GTPBP8
Gene Name
GTP binding protein 8 (putative)
There are no transcripts associated with this gene.

Gene
NEPRO
Gene Name
nucleolus and neural progenitor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319109.1 2807 UTR 3 NP_001306038.1
NM_001319110.1 2807 UTR 3 NP_001306039.1
NM_001319111.1 2807 UTR 3 NP_001306040.1
NM_001319112.1 2807 UTR 3 NP_001306041.1
NM_001319114.1 2807 UTR 3 NP_001306043.1
NM_001319115.1 2807 UTR 3 NP_001306044.1
NM_015412.3 2807 UTR 3 NP_056227.2
XM_005247276.2 2807 Intron XP_005247333.1
XM_011512635.2 2807 Intron XP_011510937.1
XM_011512636.2 2807 Intron XP_011510938.1
XM_011512638.2 2807 Intron XP_011510940.1
XM_017006102.1 2807 UTR 3 XP_016861591.1
XM_017006103.1 2807 Intron XP_016861592.1

View Full Product Details