Product Details

SNP ID
rs147494762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42658658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATTGCTGCAGATGGCTGACAT[C/T]GCTGCGTCCTGCCAAGAGCTGCTGG
Phenotype
MIM: 161565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928323 PubMed Links

Gene Details

Gene
LOC101928323
Gene Name
uncharacterized LOC101928323
There are no transcripts associated with this gene.

Gene
NKTR
Gene Name
natural killer cell triggering receptor
There are no transcripts associated with this gene.

Gene
ZBTB47
Gene Name
zinc finger and BTB domain containing 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145166.3 584 Silent Mutation ATC,ATT I101I NP_660149.2

View Full Product Details