Product Details

SNP ID
rs148262628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46579340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATTCGGCCTCGGTCTTCCCAGC[A/G]TGTGCCGCCCATGGGGATACAGCAC
Phenotype
MIM: 607180 MIM: 187395
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC2 PubMed Links

Gene Details

Gene
LRRC2
Gene Name
leucine rich repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024512.4 Intron NP_078788.2
XM_011534110.2 Intron XP_011532412.1
XM_017007177.1 Intron XP_016862666.1
Gene
TDGF1
Gene Name
teratocarcinoma-derived growth factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174136.1 Intron NP_001167607.1
NM_003212.3 Intron NP_003203.1

View Full Product Details