Product Details
- SNP ID
-
rs149313297
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:19950964 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGGAAATAAAATATGCCAGTTCAA[A/G]CTAGTACTTCTGGGAGAGTCCGCTG
- Phenotype
-
MIM: 179512
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EFHB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6773437] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EFHB
- Gene Name
- EF-hand domain family member B
There are no transcripts associated with this gene.
- Gene
- RAB5A
- Gene Name
- RAB5A, member RAS oncogene family
View Full Product Details