Product Details
- SNP ID
-
rs149047307
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:40426039 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTCTGTAGTGTGGCGATCGTGGC[G/T]GTAGCTGGAGCAGCAAATGGCACGT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RBM47
PubMed Links
Gene Details
- Gene
- RBM47
- Gene Name
- RNA binding motif protein 47
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098634.1 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
NP_001092104.1 |
NM_019027.3 |
2478 |
Silent Mutation |
ACA,ACC |
T480T |
NP_061900.2 |
XM_005248103.3 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_005248160.1 |
XM_005248107.3 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_005248164.1 |
XM_005248108.3 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_005248165.1 |
XM_005248109.4 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_005248166.1 |
XM_011513704.2 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_011512006.1 |
XM_011513707.2 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_011512009.1 |
XM_011513708.2 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_011512010.1 |
XM_017008304.1 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_016863793.1 |
XM_017008305.1 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_016863794.1 |
XM_017008306.1 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_016863795.1 |
XM_017008307.1 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_016863796.1 |
XM_017008308.1 |
2478 |
Silent Mutation |
ACA,ACC |
T549T |
XP_016863797.1 |
XM_017008309.1 |
2478 |
Silent Mutation |
ACA,ACC |
T548T |
XP_016863798.1 |
XM_017008310.1 |
2478 |
Silent Mutation |
ACA,ACC |
T480T |
XP_016863799.1 |
View Full Product Details