Product Details

SNP ID
rs150651981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70335531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAATAACCTCTGAAGTCTCTGG[A/C]ACACTAAAGGACAGCAGTGCTGGTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CABS1 PubMed Links

Gene Details

Gene
CABS1
Gene Name
calcium binding protein, spermatid associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033122.3 578 Silent Mutation GGA,GGC G164G NP_149113.3

View Full Product Details