Product Details

SNP ID
rs151085866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1349633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCAGATTGTGGAGGAACTCTTC[G/T]TCAGGTCTCACCAGTTCCGGATGCT
Phenotype
MIM: 606801 MIM: 614632
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAEA PubMed Links

Gene Details

Gene
MAEA
Gene Name
macrophage erythroblast attacher
There are no transcripts associated with this gene.

Gene
UVSSA
Gene Name
UV stimulated scaffold protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317934.1 978 Missense Mutation GTC,TTC V70F NP_001304863.1
NM_001317935.1 978 Missense Mutation GTC,TTC V70F NP_001304864.1
NM_020894.3 978 Missense Mutation GTC,TTC V70F NP_065945.2
XM_017008490.1 978 Missense Mutation GTC,TTC V70F XP_016863979.1
XM_017008491.1 978 Missense Mutation GTC,TTC V70F XP_016863980.1
XM_017008492.1 978 Missense Mutation GTC,TTC V70F XP_016863981.1
XM_017008493.1 978 Missense Mutation GTC,TTC V70F XP_016863982.1
XM_017008494.1 978 Missense Mutation GTC,TTC V70F XP_016863983.1
XM_017008495.1 978 Intron XP_016863984.1
XM_017008496.1 978 Missense Mutation GTC,TTC V179F XP_016863985.1
XM_017008497.1 978 Missense Mutation GTC,TTC V179F XP_016863986.1
XM_017008498.1 978 Missense Mutation GTC,TTC V70F XP_016863987.1
XM_017008499.1 978 Missense Mutation GTC,TTC V70F XP_016863988.1
XM_017008500.1 978 Intron XP_016863989.1

View Full Product Details