Product Details
- SNP ID
-
rs145794748
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:132823444 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGCCGCTTGGCGCCTGGCCCACG[C/T]TTGTGCCTCCCCCTGCAGCCGCTCC
- Phenotype
-
MIM: 611179
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC107984115
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4705870] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC107984115
- Gene Name
- laforin-like
There are no transcripts associated with this gene.
- Gene
- SHROOM1
- Gene Name
- shroom family member 1
- Gene
- SOWAHA
- Gene Name
- sosondowah ankyrin repeat domain family member A
There are no transcripts associated with this gene.
View Full Product Details