Product Details

SNP ID
rs147664127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179863537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCCTTGATCTTGAGTCCAATGT[A/C]CACTCTCTTCTCAAAGAAGTTCACC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C5orf45 PubMed Links

Gene Details

Gene
C5orf45
Gene Name
chromosome 5 open reading frame 45
There are no transcripts associated with this gene.

Gene
LOC100996419
Gene Name
uncharacterized LOC100996419
There are no transcripts associated with this gene.

Gene
TBC1D9B
Gene Name
TBC1 domain family member 9B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015043.3 3650 Missense Mutation NP_055858.2
NM_198868.2 3650 Missense Mutation NP_942568.2

View Full Product Details