Product Details

SNP ID
rs139009699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89435911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTATTGTCTTTGTTTTATTTC[C/T]TCAAGGTCACATGGCTTTCCTTGGT
Phenotype
MIM: 610583
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD6 PubMed Links

Gene Details

Gene
ANKRD6
Gene Name
ankyrin repeat domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242809.1 Intron NP_001229738.1
NM_001242811.1 Intron NP_001229740.1
NM_001242813.1 Intron NP_001229742.1
NM_001242814.1 Intron NP_001229743.1
NM_014942.4 Intron NP_055757.3
XM_005248679.3 Intron XP_005248736.1
XM_005248680.3 Intron XP_005248737.1
XM_005248681.2 Intron XP_005248738.1
XM_005248682.3 Intron XP_005248739.1
XM_005248683.3 Intron XP_005248740.1
XM_005248684.3 Intron XP_005248741.1
XM_011535596.2 Intron XP_011533898.1
XM_011535597.2 Intron XP_011533899.1
XM_011535598.2 Intron XP_011533900.1
XM_011535599.2 Intron XP_011533901.1
XM_011535600.2 Intron XP_011533902.1
XM_017010493.1 Intron XP_016865982.1
XM_017010494.1 Intron XP_016865983.1
XM_017010495.1 Intron XP_016865984.1
XM_017010496.1 Intron XP_016865985.1
XM_017010497.1 Intron XP_016865986.1
XM_017010498.1 Intron XP_016865987.1
XM_017010499.1 Intron XP_016865988.1
XM_017010500.1 Intron XP_016865989.1
XM_017010501.1 Intron XP_016865990.1
XM_017010502.1 Intron XP_016865991.1
XM_017010503.1 Intron XP_016865992.1
XM_017010504.1 Intron XP_016865993.1
XM_017010505.1 Intron XP_016865994.1
XM_017010506.1 Intron XP_016865995.1
XM_017010507.1 Intron XP_016865996.1
XM_017010508.1 Intron XP_016865997.1
XM_017010509.1 Intron XP_016865998.1
XM_017010510.1 Intron XP_016865999.1
XM_017010511.1 Intron XP_016866000.1
XM_017010512.1 Intron XP_016866001.1
XM_017010513.1 Intron XP_016866002.1
XM_017010514.1 Intron XP_016866003.1
XM_017010515.1 Intron XP_016866004.1

View Full Product Details