Product Details

SNP ID
rs145333192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31721672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGACAGGGTGAGCAGCATAAGG[A/G]CTTTCATGGCGAGGGTCCTGAGAAT
Phenotype
MIM: 606520 MIM: 602872 MIM: 604744 MIM: 610435 MIM: 606038 MIM: 610437
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf25 PubMed Links

Gene Details

Gene
C6orf25
Gene Name
chromosome 6 open reading frame 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025260.3 63 Intron NP_079536.2
NM_138272.2 63 Intron NP_612116.1
NM_138273.2 63 Intron NP_612117.1
NM_138274.2 63 Intron NP_612118.1
NM_138275.2 63 Intron NP_612119.1
NM_138277.2 63 Intron NP_612121.1
XM_011514920.2 63 Intron XP_011513222.1
XM_011514921.2 63 Intron XP_011513223.1
XM_017011330.1 63 Intron XP_016866819.1
XM_017011331.1 63 Intron XP_016866820.1
XM_017011332.1 63 Intron XP_016866821.1
XM_017011333.1 63 Intron XP_016866822.1
Gene
CLIC1
Gene Name
chloride intracellular channel 1
There are no transcripts associated with this gene.

Gene
DDAH2
Gene Name
dimethylarginine dimethylaminohydrolase 2
There are no transcripts associated with this gene.

Gene
LY6G6C
Gene Name
lymphocyte antigen 6 complex, locus G6C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025261.2 63 Missense Mutation GCC,GTC A3V NP_079537.1
Gene
LY6G6D
Gene Name
lymphocyte antigen 6 complex, locus G6D
There are no transcripts associated with this gene.

Gene
LY6G6E
Gene Name
lymphocyte antigen 6 complex, locus G6E (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details