Product Details

SNP ID
rs140207092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:156643238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCAGTATCTTGCGCAGCAAAGC[A/G]GTAGAGTTCAATTGGAACTCAAGTT
Phenotype
MIM: 610242 MIM: 610241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf13 PubMed Links

Gene Details

Gene
C7orf13
Gene Name
chromosome 7 open reading frame 13
There are no transcripts associated with this gene.

Gene
RNF32
Gene Name
ring finger protein 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184996.1 Intron NP_001171925.1
NM_001184997.1 Intron NP_001171926.1
NM_001308273.1 Intron NP_001295202.1
NM_001308274.1 Intron NP_001295203.1
NM_030936.3 Intron NP_112198.1
XM_005249522.4 Intron XP_005249579.1
XM_011515804.2 Intron XP_011514106.1
XM_011515805.2 Intron XP_011514107.1
XM_011515806.2 Intron XP_011514108.1
XM_011515807.2 Intron XP_011514109.1
XM_011515808.2 Intron XP_011514110.1
XM_011515809.2 Intron XP_011514111.1
XM_011515810.2 Intron XP_011514112.1
XM_011515811.2 Intron XP_011514113.1
XM_011515812.2 Intron XP_011514114.1
XM_011515813.2 Intron XP_011514115.1
XM_017011753.1 Intron XP_016867242.1
XM_017011754.1 Intron XP_016867243.1
XM_017011755.1 Intron XP_016867244.1

View Full Product Details