Product Details

SNP ID
rs142273025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:5063895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATGTGGAAAGAATTTAGAATCT[A/T]TTTCGCAATTAATTAGTAGTGATGG
Phenotype
MIM: 608191
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RBAK PubMed Links

Gene Details

Gene
RBAK
Gene Name
RB associated KRAB zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204456.1 763 Missense Mutation ATT,TTT I147F NP_001191385.1
NM_021163.3 763 Missense Mutation ATT,TTT I147F NP_066986.1
Gene
RBAK-RBAKDN
Gene Name
RBAK-RBAKDN readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204513.1 763 Intron NP_001191442.1
Gene
RBAKDN
Gene Name
RBAK downstream neighbor (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details