Product Details

SNP ID
rs142396932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:148591029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTGCCCAGTGGGGCAAGGCGCC[A/G]GATTGACCTTCGCCTGAGTGGGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf33 PubMed Links

Gene Details

Gene
C7orf33
Gene Name
chromosome 7 open reading frame 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145304.3 465 Missense Mutation CAG,CGG Q35R NP_660347.1

View Full Product Details