Product Details

SNP ID
rs146034331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99547988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCTCTCATCGAGTTGGATTGC[A/G]AAGTCTATACCGGACTGCAGCCGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM200A PubMed Links

Gene Details

Gene
FAM200A
Gene Name
family with sequence similarity 200 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145111.3 604 Silent Mutation TTC,TTT F140F NP_659802.1
XM_011515911.2 604 Silent Mutation TTC,TTT F140F XP_011514213.1
XM_011515913.2 604 Silent Mutation TTC,TTT F140F XP_011514215.1
Gene
ZNF655
Gene Name
zinc finger protein 655
There are no transcripts associated with this gene.

View Full Product Details